missing translation for 'onlineSavingsMsg'
Learn More

Sodium Phosphate, Monobasic, Monohydrate, MP Biomedicals™

Product Code. 11434662 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11434662 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11434662 Supplier MP Biomedicals Supplier No. 02191442.5
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Widely used in molecular biology, biochemistry and chromatography. Sodium Phosphate, Monobasic, Monohydrate, MP Biomedicals is a reagent with very high buffering capacity. It is is useful in conjunction with Sodium Phosphate, Dibasic in the preparation of

Sodium phosphate occurs in several forms: monobasic (NaH2PO4), dibasic (Na2HPO4), and tribasic (Na3PO4). Most neutral sodium phosphate buffer solutions consist of mixtures of the monobasic and dibasic forms to varying degrees, depending on the desired pH for the application. IIt is used in many applications, including the purification of antibodies.

Specifications

CAS 10049-21-5
Physical Form Powder
Quantity 500 g
Molecular Formula H5NaO5P
MDL Number MFCD00149208
Synonym sodium dihydrogen phosphate monohydrate, sodium phosphate monobasic monohydrate, monosodium phosphate monohydrate, unii-593yog76rn, phosphoric acid, monosodium salt, monohydrate, sodium phosphate monobasic hydrate, sodium dihydrogen phosphate hydrate, sodium dihydrogenphosphate monohydrate, sodium hydrate dihydrogen phosphate, pubchem12707
Solubility Information Freely soluble in water; insoluble in ethanol.
InChI Key BBMHARZCALWXSL-UHFFFAOYSA-N
SMILES O.[Na+].OP(O)(O)=O
IUPAC Name sodium phosphoric acid hydrate
Molecular Weight (g/mol) 139.00
PubChem CID 516949
Formula Weight 137.99
Percent Purity 98.0% to 102.0%
Grade ACS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.