missing translation for 'onlineSavingsMsg'
Learn More

Sodium phosphate monobasic monohydrate, Puriss. p.a., ACS reagent, ≥99.0% (T), Solstice

Product Code. 16655913
Change view
Click to view available options
25 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
16655913 25 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16655913 Supplier Fluka Supplier No. 7150425KG
  • ONLINE EXCLUSIVE PRICE
    2045.00 €
    1846.05 € / 25kg
    Save 198.95 €
    9% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Melting Point 650°C (anhydrous substance)
CAS 10049-21-5
Color White
CAS Max % 1
pH 4.1 to 4.5
Quantity 25 kg
Molecular Formula H5NaO5P
Linear Formula NaH2PO4 · H2O
MDL Number MFCD00149208
Synonym Monosodium phosphate; Sodium dihydrogen phosphate monohydrate
InChI Key BBMHARZCALWXSL-UHFFFAOYSA-N
SMILES O.[Na+].OP(O)(O)=O
IUPAC Name sodium phosphoric acid hydrate
Molecular Weight (g/mol) 139.00
Grade ACS
Chemical Name or Material Sodium phosphate monobasic monohydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.