missing translation for 'onlineSavingsMsg'
Learn More

Sodium phosphate, tribasic, anhydrous, tech.

Product Code. 11326778
Click to view available options
Quantity:
1 kg
5 kg
Unit Size:
1kg
5kg
This item is not returnable. View return policy

Product Code. 11326778

missing translation for 'mfr': Thermo Scientific Alfa Aesar 013438.A1

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 7601-54-9 | Na3O4P | 163.939 g/mol

Sodium phosphate, tribasic is used in soaps, detergents, photographic developers, water softeners and boiler scale removers. It is used as a food additive, as cleaning agents, as a protein precipitant and an active ingredient in fluxes designed to deoxygenate nonferrous metals for casting. It is used to prepare chlorinated trisodium phosphate by reacting with sodium hypochlorite and used as a disinfectant and bleach. Further, it is used in dairy substitutes and to inhibit mold. In addition, it is employed in ceramic production. It is also used for the preservation of biologically active soil samples and fractionization of nucleotides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7601-54-9
Molecular Formula Na3O4P
Molecular Weight (g/mol) 163.939
MDL Number MFCD00003510
InChI Key RYFMWSXOAZQYPI-UHFFFAOYSA-K
Synonym trisodium phosphate, sodium orthophosphate, tromete, tribasic sodium phosphate, phosphoric acid, trisodium salt, oakite, sodium phosphate, anhydrous, trisodium orthophosphate, phosphoric acid trisodium salt, trinatriumphosphat
PubChem CID 24243
ChEBI CHEBI:37583
IUPAC Name trisodium;phosphate
SMILES [O-]P(=O)([O-])[O-].[Na+].[Na+].[Na+]

Specifications

Color White
Physical Form Powder
Quantity 1 kg
Solubility Information Soluble in water. Insoluble in ethanol and carbon disulfide.
Formula Weight 163.94
Grade Technical
Odor Odorless
Sensitivity Hygroscopic
Appearance White powder
Density 1.62g/cm3
Chemical Name or Material Sodium phosphate, tribasic, anhydrous
Show More Show Less
compliance-icons
Product Identifier
  • Sodium phosphate, tribasic, anhydrous
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : trisodium phosphate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.