missing translation for 'onlineSavingsMsg'
Learn More

Sodium phosphate tribasic, Puriss., anhydrous, ≥94%, Honeywell

Product Code. 15620760 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
40 kg
Packaging:
Fiberboard Box
Unit Size:
40kg
Product Code. Quantity Packaging unitSize
15620760 40 kg Fiberboard Box 40kg
1 options
This item is not returnable. View return policy

Product Code. 15620760

Brand: Honeywell Research Chemicals 0427840KG

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 7601-54-9
Physical Form Anhydrous
Quantity 40 kg
Molecular Formula Na3O4P
Linear Formula Na3PO4
MDL Number MFCD00003510
UN Number NONH for all modes of transport
Synonym trisodium phosphate, sodium orthophosphate, tromete, tribasic sodium phosphate, phosphoric acid, trisodium salt, oakite, sodium phosphate, anhydrous, trisodium orthophosphate, phosphoric acid trisodium salt, trinatriumphosphat
InChI Key RYFMWSXOAZQYPI-UHFFFAOYSA-K
SMILES [O-]P(=O)([O-])[O-].[Na+].[Na+].[Na+]
IUPAC Name trisodium;phosphate
Molecular Weight (g/mol) 163.939
PubChem CID 24243
ChEBI CHEBI:37583
Formula Weight 163.94g/mol
Percent Purity >94%
Grade puriss.
Packaging Fiberboard Box
Chemical Name or Material Sodium phosphate tribasic, Puriss., anhydrous, ≥94%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.