missing translation for 'onlineSavingsMsg'
Learn More

Sodium phosphonoformate hexahydrate, 98+%

Product Code. 11466526
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11466526 25 g 25g
11446526 1 g 1g
11456526 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 11466526 Leverandør Thermo Scientific Alfa Aesar Leverandørnr. B24784.14
 Restricted Item
Estimated Shipment: 02-12-2026
Læg i indkøbskurv
Læg i indkøbskurv
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy

CAS: 34156-56-4 | CH12Na3O11P | 300.04 g/mol

Sodium phosphonoformate hexahydrate acts as an antiviral agent and reverse transcriptase inhibitor. It is also used to inhibit viral DNA polymerase. Further, it is used as a type II Pi transporter inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium phosphonoformate hexahydrate acts as an antiviral agent and reverse transcriptase inhibitor. It is also used to inhibit viral DNA polymerase. Further, it is used as a type II Pi transporter inhibitor.

Solubility
Soluble in water.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 34156-56-4
Molecular Formula CH12Na3O11P
Molecular Weight (g/mol) 300.04
MDL Number MFCD00150176
InChI Key ILRVASBWNRYBFD-UHFFFAOYSA-K
Synonym foscarnet sodium hexahydrate, sodium phosphonatoformate hexahydrate, unii-964ys0oog1, foscarnet trisodium hexahydrate, phosphonoformic acid trisodium salt hexahydrate, trisodium phosphonoformte hexahydrate, sodium phosphonoformate tribasic hexahydrate, phosphonoformic acid, trisodium salt, hexahydrate, trisodium foscarnet 3-hexahydrate, formic acid, phosphono-, trisodium salt, hexahydrate
PubChem CID 169569
ChEBI CHEBI:60269
SMILES O.O.O.O.O.O.[Na+].[Na+].[Na+].[O-]C(=O)P([O-])([O-])=O

Tekniske data

Quantity 25 g
Solubility Information Soluble in water.
IUPAC Name trisodium hexahydrate phosphonatoformate
Formula Weight 300.08 (191.98 Anhydrous)
Percent Purity ≥98%
Chemical Name or Material Sodium phosphonoformate hexahydrate

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.