missing translation for 'onlineSavingsMsg'
Learn More

Sodium polyphosphate, pure

Product Code. 10025533 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Plastic bottle
Unit Size:
1kg
250g
5kg
This item is not returnable. View return policy

Product Code. 10025533

Brand: Thermo Scientific Acros 390930050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 50813-16-6 | (NaO3P)n

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50813-16-6
Molecular Formula (NaO3P)n
MDL Number MFCD00134118
Synonym hexasodium oxido oxido phosphonatooxy phosphoryl oxy phosphoryl oxy phospho phosphonato oxy phosphinate
IUPAC Name Sodium polyphosphate
SMILES [Na]OP(-*)(=O)O-*

Specifications

Melting Point 600.0°C
Color White
Physical Form Crystalline Powder
Quantity 5 kg
Linear Formula (NaPO3)n
Merck Index 15, 8773
Solubility Information Solubility in water: 1000g/l (20°C)
Percent Purity 65 to 70%
Grade Pure
Packaging Plastic bottle
Chemical Name or Material Sodium polyphosphate
Show More Show Less
compliance-icons
Product Identifier
  • Sodium polyphosphate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.