missing translation for 'onlineSavingsMsg'
Learn More

Sodium polyphosphate, pure

Product Code. 10055293 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Plastic bottle
Unit Size:
1kg
250g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10055293 250 g Plastic bottle 250g
10268893 1 kg Plastic bottle 1kg
10025533 5 kg Plastic bottle 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10055293 Supplier Thermo Scientific Acros Supplier No. 390932500
  • ONLINE EXCLUSIVE PRICE
    31.85 €
    29.90 € / 250g
    Save 1.95 €
    6% Off
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 50813-16-6 ∣ (NaO3P)n

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50813-16-6
Molecular Formula (NaO3P)n
MDL Number MFCD00134118
Synonym hexasodium oxido oxido phosphonatooxy phosphoryl oxy phosphoryl oxy phospho phosphonato oxy phosphinate
IUPAC Name Sodium polyphosphate
SMILES [Na]OP(-*)(=O)O-*

Specifications

Melting Point 600.0°C
Color White
Physical Form Crystalline Powder
Quantity 250 g
Packaging Plastic bottle
Linear Formula (NaPO3)n
Merck Index 15, 8773
Solubility Information Solubility in water: 1000g/l (20°C)
Percent Purity 65 to 70%
Grade Pure
Chemical Name or Material Sodium polyphosphate
Show More Show Less
compliance-icons
Product Identifier
  • Sodium polyphosphate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.