missing translation for 'onlineSavingsMsg'
Learn More

Sodium propionate, 99%

Product Code. p-7023652
Click to view available options
Quantity:
500 g
2500 g
Unit Size:
2500g
500g
This item is not returnable. View return policy

Product Code. 11468027

Brand: Thermo Scientific Alfa Aesar A17440.36

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 137-40-6 | C3H5NaO2 | 96.061 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Sodium propionate is used as a food preservative, as a mold inhibitor in bakery products and in insect cell culture. It is also used in pharmaceuticals, cosmetics, agriculture and animal feed as preservative. It acts as a food additive. Further, it inhibits the growth of lactic acid bacteria.

Solubility
Soluble in water. Slightly soluble in alcohol.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 137-40-6
Molecular Formula C3H5NaO2
Molecular Weight (g/mol) 96.061
MDL Number MFCD00002759
InChI Key JXKPEJDQGNYQSM-UHFFFAOYSA-M
Synonym sodium propionate, sodium propanoate, propionic acid sodium salt, propanoic acid, sodium salt, napropion, ocuseptine, deketon, impedex, keenate, propiofar
PubChem CID 2723816
IUPAC Name sodium;propanoate
SMILES CCC(=O)[O-].[Na+]

Specifications

Melting Point 285°C to 291°C
Assay Percent Range 99%
Linear Formula CH3CH2CO2Na
Quantity 500 g
Sensitivity Hygroscopic
Merck Index 14,8669
Solubility Information Soluble in water. Slightly soluble in alcohol.
Formula Weight 96.06
Percent Purity 99%
Chemical Name or Material Sodium propionate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.