missing translation for 'onlineSavingsMsg'
Learn More

Sodium Propionate, Extra Pure, SLR, Fisher Chemical

Product Code. 10182680 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Plastic powder jar
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10182680 500 g Plastic powder jar 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10182680 Supplier Fisher Chemical Supplier No. S/6100/53
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Melting Point 285°C
CAS 137-40-6
Color White
pH 8 to 10.5
Physical Form Powder/Solid
Quantity 500 g
Molecular Formula C3H5NaO2
MDL Number 2759
Synonym sodium propionate, sodium propanoate, propionic acid sodium salt, propanoic acid, sodium salt, napropion, ocuseptine, deketon, impedex, keenate, propiofar
InChI Key JXKPEJDQGNYQSM-UHFFFAOYSA-M
SMILES CCC(=O)[O-].[Na+]
IUPAC Name sodium;propanoate
Molecular Weight (g/mol) 96.061
PubChem CID 2723816
Formula Weight 96.06g/mol
Packaging Plastic powder jar
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.