missing translation for 'onlineSavingsMsg'
Learn More

Sodium Propionate, TRC

Product Code. 30413203
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30413203 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30413203 Supplier Toronto Research Chemicals Supplier No. TRCS6670655G
 Restricted Item
Estimated Shipment: 23-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 137-40-6
Molecular Formula C3 H5 O2 . Na
Molecular Weight (g/mol) 96.06
Synonym Sodium propionate, Sodium propanoate, Propionic acid sodium salt, Propanoic acid, sodium salt (1:1) (ACI), Propanoic acid, sodium salt (9CI), Propionic acid, sodium salt (8CI), Bioban-S, Deketon, Impedex, Mycoban, Napropion, Ocuseptine, Propi-Ophtal, Propiofar, Propion, Propisol, Sodium ethanecarboxylate, Whit-Pro
IUPAC Name sodium;propanoate
SMILES [Na+].CCC(=O)[O-]

Specifications

Quantity 5 g
InChI Formula InChI=1S/C3H6O2.Na/c1-2-3(4)5;/h2H2,1H3,(H,4,5);/q;+1/p-1
Formula Weight 96.0187
Chemical Name or Material Propionic acid sodium salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.