missing translation for 'onlineSavingsMsg'
Learn More

Sodium Propylparaben, TRC

Product Code. 30136074
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136074 5 g 5g
30133066 25 g 25g
30005413 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136074 Supplier Toronto Research Chemicals Supplier No. TRCS6732655G
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35285-69-9
Molecular Formula C10 H11 O3 . Na
Molecular Weight (g/mol) 202.18
Synonym Benzoic acid, 4-hydroxy-, propyl ester, sodium salt (9CI), Nipasol M Sodium, Propyl 4-hydroxybenzoate sodium salt, Propyl p-hydroxybenzoate sodium salt, Propylparaben sodium, Sodium 4-propoxycarbonylphenoxide, Sodium propyl 4-hydroxybenzoate, Sodium propyl p-hydroxybenzoate, Sodium propyl paraben
IUPAC Name sodium;4-propoxycarbonylphenolate
SMILES [Na+].CCCOC(=O)c1ccc([O-])cc1

Specifications

Quantity 5 g
InChI Formula InChI=1S/C10H12O3.Na/c1-2-7-13-10(12)8-3-5-9(11)6-4-8;/h3-6,11H,2,7H2,1H3;/q;+1/p-1
Formula Weight 202.0606
Percent Purity >95%
Chemical Name or Material Sodium Propyl Parahydroxybenzoate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.