missing translation for 'onlineSavingsMsg'
Learn More

Sodium Pyrophosphate, TRC

Product Code. 30410121
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30410121 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30410121 Supplier Toronto Research Chemicals Supplier No. TRCS66708025G
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sodium Pyrophosphate
CAS 7722-88-5
Quantity 25 g
Molecular Formula 4 Na . O7 P2
Synonym Diphosphoric acid','sodium salt (1:4), Diphosphoric acid','tetrasodium salt (9CI), Pyrophosphoric acid','tetrasodium salt (8CI), Accoline 126, Anhydrous tetrasodium pyrophosphate, Nankarin S 1, Phosphomix 110, Phosphotex, SPP, SPP (phosphate), Sodium diphosphate, Sodium diphosphate (Na4P2O7), Sodium phosphate (Na4P2O7), Sodium phosphorus oxide (Na4P2O7), Sodium pyrophosphate (Na4P2O7), TSPP, Tetrasodium diphosphate, Tetrasodium diphosphate (Na4P2O7), Tetrasodium pyrophosphate, Tetrasodium pyrophosphate (Na4P2O7), Thermphos Pyro E 450, Victor TSPP
InChI Formula InChI=1S/4Na.H4O7P2/c;;;;1-8(2,3)7-9(4,5)6/h;;;;(H2,1,2,3)(H2,4,5,6)/q4*+1;/p-4
SMILES [Na+].[Na+].[Na+].[Na+].[O-]P(=O)([O-])OP(=O)([O-])[O-]
IUPAC Name tetrasodium;phosphonato phosphate
Molecular Weight (g/mol) 265.9024
Formula Weight 265.871
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.