missing translation for 'onlineSavingsMsg'
Learn More

Sodium Pyruvate, BAKER ANALYZED™ Reagent, J.T. Baker™

Product Code. 15588114 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
1 kg
Packaging:
Wide Mouth Amber Glass Bottle
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15588114 1 kg Wide Mouth Amber Glass Bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15588114 Supplier Avantor J.T.Baker Supplier No. 335402
  • ONLINE EXCLUSIVE PRICE
    1166.18 €
    1048.32 € / 1kg
    Save 117.86 €
    10% Off
 Restricted Item
Estimated Shipment: 22-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High quality chemicals for laboratory and specialized industrial use

Kemiske identifikatorer

CAS 113-24-6
Molecular Formula C3H3NaO3
Molecular Weight (g/mol) 110.044
InChI Key DAEPDZWVDSPTHF-UHFFFAOYSA-M
Synonym sodium pyruvate, pyruvic acid sodium salt, sodium 2-oxopropanoate, pyruvic acid, sodium salt, propanoic acid, 2-oxo-, sodium salt, sodium alpha-ketopropionate, pyruvate sodium, unii-pod38aif08, 2-oxopropanoic acid sodium salt, pod38aif08
PubChem CID 23662274
ChEBI CHEBI:50144
IUPAC Name sodium;2-oxopropanoate
SMILES CC(=O)C(=O)[O-].[Na+]

Tekniske data

Quantity 1 kg
Formula Weight 110.05
Percent Purity ≥99.6%
Grade BAKER ANALYZED™ Reagent
Packaging Wide Mouth Amber Glass Bottle
Chemical Name or Material Sodium Pyruvate
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.