missing translation for 'onlineSavingsMsg'
Learn More

Sodium pyruvate, MP Biomedicals™

Product Code. 11440465 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11440465 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11440465 Supplier MP Biomedicals Supplier No. 0219965480
 Restricted Item
Estimated Shipment: 15-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Sodium pyruvate, White Powder

  • It is utilized as a component in culture broth and media.
  • The use of sodium pyruvate in Wallen fermentation medium to enhance the conversion of oleic acid to 10-ketostearic acid by Bacillus sphaericus has been described.
  • A protocol that uses sodium pyruvate to establish stably transfected human B cell lines has been published.

Chemical Identifiers

CAS 113-24-6
Molecular Formula C3H3NaO3
Molecular Weight (g/mol) 110.04
MDL Number MFCD00002586
InChI Key DAEPDZWVDSPTHF-UHFFFAOYSA-M
Synonym Pyruvic acid sodium salt, 2-Oxopropanoic acid sodium salt, α-ketopropionic acid sodium salt

Specifications

Melting Point >300°C (literature)
Color White
pH 6.54
Loss on Drying 0.0021
Quantity 100 g
Beilstein 3568341
Solubility Information Soluble in water (0.06g/mL) - Clear and Complete.
Physical Form Powder
Assay ≥98.0%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.