missing translation for 'onlineSavingsMsg'
Learn More

Sodium Salicylate, 99%, Thermo Scientific Chemicals

Catalog Number p-7023470
Click to view available options
Quantity:
250 g
1000 g
54-21-7
C7H5NaO3
160.104
MFCD00002440
ABBQHOQBGMUPJH-UHFFFAOYSA-M
sodium salicylate, sodium 2-hydroxybenzoate, salsonin, clin, enterosalicyl, enterosalil, entrosalyl, glutosalyl, kerasalicyl, magsalyl
16760658
CHEBI:9180
sodium;2-hydroxybenzoate
C1=CC=C(C(=C1)C(=O)[O-])O.[Na+]
54-21-7
C7H5NaO3
160.104
MFCD00002440
ABBQHOQBGMUPJH-UHFFFAOYSA-M
sodium salicylate, sodium 2-hydroxybenzoate, salsonin, clin, enterosalicyl, enterosalil, entrosalyl, glutosalyl, kerasalicyl, magsalyl
16760658
CHEBI:9180
sodium;2-hydroxybenzoate
C1=CC=C(C(=C1)C(=O)[O-])O.[Na+]

Sodium salicylate is used as an analgesic and antipyretic. It also acts as a non-steroidal anti-inflammatory drug (NSAID). Further, it is used as a phosphor for the detection of vacuum ultraviolet radiation and electrons. It serves as a neurofibromatosis kappa B inhibitor and reducer of oxidative stress. It finds applications as a drug with the function of being antifebrile and anti-rheumatic.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium salicylate is used as an analgesic and antipyretic. It also acts as a non-steroidal anti-inflammatory drug (NSAID). Further, it is used as a phosphor for the detection of vacuum ultraviolet radiation and electrons. It serves as a neurofibromatosis kappa B inhibitor and reducer of oxidative stress. It finds applications as a drug with the function of being antifebrile and anti-rheumatic.

Solubility
Soluble in water, 1,4-dioxane, ethanol and glycerol.

Notes
Light sensitive. Store in a cool place. Incompatible with strong oxidizing agents, strong acidsmineral acids, metallic salts and iodine.
TRUSTED_SUSTAINABILITY
Melting Point >300°C
Odor Odorless
Quantity 250 g
Beilstein 3732792
Sensitivity Light sensitive
Merck Index 14,8332
Solubility Information Soluble in water,1,4-dioxane,ethanol and glycerol.
Formula Weight 160.11
Percent Purity 99%
Chemical Name or Material Sodium salicylate
compliance-icons
Product Identifier
  • Sodium salicylate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'