missing translation for 'onlineSavingsMsg'
Learn More

Sodium Salicylate, Extra Pure, SLR, Fisher Chemical™

Product Code. 10030460 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Plastic powder jar
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10030460 500 g Plastic powder jar 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10030460 Supplier Fisher Chemical Supplier No. S/6200/53
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

C7H5NaO3, CAS Number-54-21-7, clin, enterosalicyl, enterosalil, entrosalyl, glutosalyl, kerasalicyl, magsalyl, salsonin, sodium 2-hydroxybenzoate, sodium salicylate, 500g, CHEBI:9180, White, 160.1g/mol, ABBQHOQBGMUPJH-UHFFFAOYSA-M, sodium;2-hydroxybenzoate, 200 deg.C, 2440, 160.104 | CAS: 54-21-7 | C7H5NaO3 | 160.104 g/mol

C7H5NaO3, CAS Number-54-21-7, clin, enterosalicyl, enterosalil, entrosalyl, glutosalyl, kerasalicyl, magsalyl, salsonin, sodium 2-hydroxybenzoate, sodium salicylate, 500g, CHEBI:9180, White, 160.1g/mol, ABBQHOQBGMUPJH-UHFFFAOYSA-M, sodium;2-hydroxybenzoate, 200 deg.C, 2440, 160.104

TRUSTED_SUSTAINABILITY

Specifications

Melting Point 200°C
CAS 54-21-7
Color White
Physical Form Solid
Quantity 500 g
Molecular Formula C7H5NaO3
MDL Number 2440
Synonym sodium salicylate, sodium 2-hydroxybenzoate, salsonin, clin, enterosalicyl, enterosalil, entrosalyl, glutosalyl, kerasalicyl, magsalyl
InChI Key ABBQHOQBGMUPJH-UHFFFAOYSA-M
SMILES C1=CC=C(C(=C1)C(=O)[O-])O.[Na+]
IUPAC Name sodium;2-hydroxybenzoate
Molecular Weight (g/mol) 160.104
PubChem CID 16760658
ChEBI CHEBI:9180
Formula Weight 160.1g/mol
Grade Extra Pure
Packaging Plastic powder jar
Show More Show Less
compliance-icons
Product Identifier
  • Salicylic acid, sodium salt
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash face, hands and any thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.