missing translation for 'onlineSavingsMsg'
Learn More

Sodium Selenate, ≥95% (elemental analysis), Honeywell™ Fluka™

Product Code. 15668070 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
10 g
100 g
Packaging:
Poly bottle
Unit Size:
100g
10g
Product Code. Quantity Packaging unitSize
15668070 10 g Poly bottle 10g
15658070 100 g Poly bottle 100g
2 options
This item is not returnable. View return policy

Product Code. 15668070

Brand: Honeywell-Fluka S088210G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

≥95% (elemental analysis)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13410-01-0
Molecular Formula Na2O4Se
Molecular Weight (g/mol) 188.95
MDL Number MFCD00003490
InChI Key MHQOTKLEMKRJIR-UHFFFAOYSA-L
Synonym sodium selenate, disodium selenate, natriumseleniat, selenic acid, disodium salt, caswell no. 791, natriumseleniat german, sel-tox sso2 and ss-20, unii-5dqp25600a, selenic acid h2seo4 , disodium salt, ccris 1259
PubChem CID 25960
ChEBI CHEBI:77775
SMILES [Na+].[Na+].[O-][Se]([O-])(=O)=O

Specifications

Color White
pH 5.5 to 7.5 (at 18.90g/L 25°C)
Quantity 10 g
Linear Formula Na2SeO4
UN Number UN2630
Solubility Information Soluble in water
IUPAC Name disodium;selenate
Formula Weight 188.94g/mol
Percent Purity ≥95% (elemental analysis)
Odor Odorless
Packaging Poly bottle
Appearance Crystalline
Density 3.213g/cu.cm at 17.5°C
Chemical Name or Material Sodium selenate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.