missing translation for 'onlineSavingsMsg'
Learn More

Sodium selenate, anhydrous, 99.8+% (metals basis)

Product Code. 11326459
Change view
Click to view available options
Quantity:
5 g
50 g
250 g
1 kg
Unit Size:
1kg
250g
50g
5g
Product Code. Quantity unitSize
11326459 50 g 50g
11356459 5 g 5g
11336459 250 g 250g
11346459 1 kg 1kg
4 options
This item is not returnable. View return policy

Product Code. 11326459

Brand: Thermo Scientific Alfa Aesar 012613.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For Use As a Reagent | CAS: 13410-01-0 | Na2O4Se | 188.95 g/mol

Sodium selenate is used in glass industry, and produces red hue in glass. It acts as a decoloring agent in glass production. Combined with ferrous oxides, it produces colorless glass. Appropriate preparation of sodium selenate is used in multivitamins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13410-01-0
Molecular Formula Na2O4Se
Molecular Weight (g/mol) 188.95
MDL Number MFCD00003490
InChI Key MHQOTKLEMKRJIR-UHFFFAOYSA-L
Synonym sodium selenate, disodium selenate, natriumseleniat, selenic acid, disodium salt, caswell no. 791, natriumseleniat german, sel-tox sso2 and ss-20, unii-5dqp25600a, selenic acid h2seo4 , disodium salt, ccris 1259
PubChem CID 25960
ChEBI CHEBI:77775
IUPAC Name disodium;selenate
SMILES [Na+].[Na+].[O-][Se]([O-])(=O)=O

Specifications

Melting Point 32° (transition)
Color Purple
Physical Form Powder
Quantity 50 g
Assay Percent Range 99.8+% (metals basis)
Solubility Information Soluble in water.
Formula Weight 188.94
Odor Odorless
Sensitivity Hygroscopic
Appearance White powder
Density 3.213
Chemical Name or Material Sodium selenate, anhydrous
Show More Show Less
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H300-Fatal if swallowed.
  • H315-Causes skin irritation.
  • H330-Fatal if inhaled.
  • H373-May cause damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.