missing translation for 'onlineSavingsMsg'
Learn More

Sodium Stearate, TRC

Product Code. 30414087
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30414087 10 g 10g
30412358 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30414087 Supplier Toronto Research Chemicals Supplier No. TRCS96005510G
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sodium Stearate
CAS 822-16-2
Molecular Formula C18H35NaO2
Quantity 10 g
Synonym Sodium Stearate
InChI Formula InChI=1S/C18H36O2.Na/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20;/h2-17H2,1H3,(H,19,20);/q;+1/p-1
SMILES [Na+].CCCCCCCCCCCCCCCCCC(=O)[O-]
IUPAC Name sodium;octadecanoate
Molecular Weight (g/mol) 306.46
Formula Weight 306.2535
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.