missing translation for 'onlineSavingsMsg'
Learn More

Sodium sulfate, ACS, 99.0% min

Product Code. 11387277
Change view
Click to view available options
Quantity:
500 g
2 kg
10 kg
Unit Size:
10kg
2kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11387277 500 g 500g
11397277 2 kg 2kg
11377277 10 kg 10kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11387277 Supplier Thermo Scientific Alfa Aesar Supplier No. 011560.36
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For Use in Drying organic Liquids, in Kjeldahl Nitrogen Determination, in Manufacturing of Glass ∣ CAS: 7757-82-6 ∣ Na2O4S ∣ 142.04 g/mol

In drying organic liquids, in Kjeldahl nitrogen determination, in manufacturing of glass. Sodium sulfate anhydrous is used primarily for drying non-aqueous solvents by removing traces of water. It is used as a fining agent for molten glass, levelling agent in textile processing, and filler in detergents. It is also used in Kraft process for paper pulping, defrosting windows, manufacture of starch, glass, and soap.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7757-82-6
Molecular Formula Na2O4S
Molecular Weight (g/mol) 142.04
MDL Number MFCD00003504
InChI Key PMZURENOXWZQFD-UHFFFAOYSA-L
Synonym sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1
PubChem CID 24436
ChEBI CHEBI:32149
IUPAC Name disodium;sulfate
SMILES [Na+].[Na+].[O-]S([O-])(=O)=O

Specifications

Melting Point 884°C
Physical Form Powder
Quantity 500 g
Merck Index 14,8680
Solubility Information Soluble in water; Insoluble in alcoholSoluble in water,glycerol and hydrogen iodide; Insoluble in ethanol and many organic solvents.
Formula Weight 142.04
Percent Purity ≥99.0%
Grade ACS
Odor Odorless
Refractive Index 1.484
Sensitivity Hygroscopic
Density 2.680
Chemical Name or Material Sodium sulfate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.