missing translation for 'onlineSavingsMsg'
Learn More

Sodium Sulfate Anhydrous, Certified AR for Analysis, Fisher Chemical™

Product Code. 10204070 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
3 kg
5 kg
Unit Size:
3kg
500g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10204070 5 kg 5kg
10204690 500 g 500g
10635892 3 kg 3kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10204070 Supplier Fisher Chemical Supplier No. S/6640/65
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Na2O4S, CAS Number-7757-82-6, disodium sulfate, disodium sulphate, salt cake, sodium sulfate, sodium sulfate 2:1, sodium sulfate anhydrous, sodium sulfate, anhydrous, sodium sulphate, sulfuric acid disodium salt, thenardite, 5kg, CHEBI:32149, White, 142.04g/mol, PMZURENOXWZQFD-UHFFFAOYSA-L

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7757-82-6
Molecular Formula Na2O4S
Molecular Weight (g/mol) 142.04
MDL Number MFCD00003504
InChI Key PMZURENOXWZQFD-UHFFFAOYSA-L
Synonym sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1
PubChem CID 24436
ChEBI CHEBI:32149
SMILES [Na+].[Na+].[O-]S([O-])(=O)=O

Specifications

Melting Point 884°C
Color White
pH 5 to 8
Physical Form Solid
Quantity 5 kg
IUPAC Name disodium sulfate
Formula Weight 142.04g/mol
Packaging Plastic powder jar
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.