missing translation for 'onlineSavingsMsg'
Learn More

Sodium Sulfate Anhydrous, Certified AR for Analysis, Fisher Chemical

Product Code. 10192730 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 kg
Packaging:
Plastic powder jar
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10192730 1 kg Plastic powder jar 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10192730 Supplier Fisher Chemical Supplier No. S/6640/60
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Melting Point 884°C
CAS 7757-82-6
Color White
pH 5 to 8
Physical Form Solid
Quantity 1 kg
Molecular Formula Na2O4S
MDL Number MFCD00003504
Synonym sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1
InChI Key PMZURENOXWZQFD-UHFFFAOYSA-L
SMILES [Na+].[Na+].[O-]S([O-])(=O)=O
IUPAC Name disodium sulfate
Molecular Weight (g/mol) 142.04
PubChem CID 24436
ChEBI CHEBI:32149
Formula Weight 142.04g/mol
Packaging Plastic powder jar
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.