missing translation for 'onlineSavingsMsg'
Learn More

Sodium Sulfate, Anhydrous p., Chem Lab

Product Code. 13288243
Change view
Click to view available options
Sodium Sulfate, Anhydrous p., 2.5 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
13288243 Sodium Sulfate, Anhydrous p., 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13288243 Supplier Chem-Lab Supplier No. CL00.1412.2500
  • ONLINE EXCLUSIVE PRICE
    76.00 €
    41.70 € / 2.5kg
    Save 34.30 €
    45% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Melting Point 888°C
CAS 7757-82-6
Color Colorless
Physical Form Solid
Quantity 5 kg
Assay Percent Range > 99%
Molecular Formula Na2O4S
MDL Number MFCD00003504
InChI Key PMZURENOXWZQFD-UHFFFAOYSA-L
SMILES [Na+].[Na+].[O-]S([O-])(=O)=O
IUPAC Name disodium sulfate
Molecular Weight (g/mol) 142.04
Grade Pure
Loss on Drying < 0.2%
Packaging Polyethylene
Density 2.7 g/cm3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.