missing translation for 'onlineSavingsMsg'
Learn More

Sodium Sulfate Decahydrate, 97%, Reagent grade, Honeywell™

Product Code. p-7120826 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
Unit Size:
2.5kg
500g
This item is not returnable. View return policy

Product Code. 15612690

Brand: Honeywell Research Chemicals 246980500G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Reagent Grade, 97%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7727-73-3
Molecular Formula H20Na2O14S
Molecular Weight (g/mol) 322.186
MDL Number MFCD00149189
InChI Key RSIJVJUOQBWMIM-UHFFFAOYSA-L
Synonym sodium sulfate decahydrate, glauber's salt, disodium sulfate decahydrate, sodium sulfate usp, sulfuric acid disodium salt, decahydrate, unii-0ypr65r21j, sodium sulfate na2so4 decahydrate, sodium sulphate decahydrate, sodium sulfate decahydrate na2so4.10h2o, disodium sulfate decahydrate na2so4.10h2o
PubChem CID 62649
ChEBI CHEBI:32586
IUPAC Name disodium;sulfate;decahydrate
SMILES O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Na+].[Na+]

Specifications

Melting Point 32.4°C
Color White
Quantity 500 g
Linear Formula Na2SO4 · 10H2O
UN Number NONH for all modes of transport
Formula Weight 322.2g/mol
Percent Purity 97%
Grade Reagent
Packaging Poly Bottle
Appearance Crystals or granules
Density 1.46g/mL at 25°C
Chemical Name or Material Sodium Sulfate Decahydrate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.