missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sodium sulfate decahydrate, 99%, for biochemistry

Product Code. 12645127 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
2.5kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
12645127 500 g Plastic bottle 500g
12672936 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12645127 Supplier Thermo Scientific Chemicals Supplier No. 447935000
  • ONLINE EXCLUSIVE PRICE
    47.30 €
    45.90 € / 500g
    Save 1.40 €
    2% Off
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 7727-73-3 | Na2O410H2O | 322.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7727-73-3
Molecular Formula Na2O410H2O
Molecular Weight (g/mol) 322.2
InChI Key RSIJVJUOQBWMIM-UHFFFAOYSA-L
Synonym sodium sulfate decahydrate, glauber's salt, disodium sulfate decahydrate, sodium sulfate usp, sulfuric acid disodium salt, decahydrate, unii-0ypr65r21j, sodium sulfate na2so4 decahydrate, sodium sulphate decahydrate, sodium sulfate decahydrate na2so4.10h2o, disodium sulfate decahydrate na2so4.10h2o
PubChem CID 62649
ChEBI CHEBI:32586
IUPAC Name disodium;sulfate;decahydrate
SMILES O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Na+].[Na+]

Specifications

Melting Point 884.0°C
Quantity 500 g
Assay Percent Range 99%
Linear Formula Na2SO4·10H2O
Solubility Information Clear colorless
Formula Weight 322.2
Percent Purity 99%
Grade Biochemical
Packaging Plastic bottle
Chemical Name or Material Sodium sulfate decahydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.