missing translation for 'onlineSavingsMsg'
Learn More

Sodium Sulfate Decahydrate, Extra Pure, SLR, Fisher Chemical

Product Code. 10162780 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 kg
Packaging:
Plastic powder jar
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10162780 1 kg Plastic powder jar 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10162780 Supplier Fisher Chemical Supplier No. S/6520/60

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Melting Point 884°C
CAS 7727-73-3
Color White
pH 5.2 to 9.2
Physical Form Solid
Quantity 1 kg
Molecular Formula H20Na2O14S
MDL Number 149189
Synonym sodium sulfate decahydrate, glauber's salt, disodium sulfate decahydrate, sodium sulfate usp, sulfuric acid disodium salt, decahydrate, unii-0ypr65r21j, sodium sulfate na2so4 decahydrate, sodium sulphate decahydrate, sodium sulfate decahydrate na2so4.10h2o, disodium sulfate decahydrate na2so4.10h2o
InChI Key RSIJVJUOQBWMIM-UHFFFAOYSA-L
SMILES O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Na+].[Na+]
IUPAC Name disodium;sulfate;decahydrate
Molecular Weight (g/mol) 322.186
PubChem CID 62649
ChEBI CHEBI:32586
Formula Weight 322.2g/mol
Packaging Plastic powder jar
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.