missing translation for 'onlineSavingsMsg'
Learn More

Sodium sulfate decahydrate, extra pure

Product Code. 10760775 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
25 kg
Packaging:
Fibre drum
Plastic bottle
Unit Size:
1kg
2.5kg
250g
25kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10760775 250 g Plastic bottle 250g
10128043 1 kg Plastic bottle 1kg
10403415 2.5 kg Plastic bottle 2.5kg
13498328 25 kg Fibre drum 25kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10760775 Supplier Thermo Scientific Supplier No. 125012500
  • ONLINE EXCLUSIVE PRICE
    26.90 €
    26.10 € / 250g
    Save 0.80 €
    2% Off
View all promotions
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 7727-73-3 | Na2O410H2O | 322.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7727-73-3
Molecular Formula Na2O410H2O
Molecular Weight (g/mol) 322.2
MDL Number MFCD00149189
InChI Key RSIJVJUOQBWMIM-UHFFFAOYSA-L
Synonym sodium sulfate decahydrate, glauber's salt, disodium sulfate decahydrate, sodium sulfate usp, sulfuric acid disodium salt, decahydrate, unii-0ypr65r21j, sodium sulfate na2so4 decahydrate, sodium sulphate decahydrate, sodium sulfate decahydrate na2so4.10h2o, disodium sulfate decahydrate na2so4.10h2o
PubChem CID 62649
ChEBI CHEBI:32586
IUPAC Name disodium;sulfate;decahydrate
SMILES O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Na+].[Na+]

Specifications

Melting Point 884.0°C
Color White
pH 5.2 to 9.2 (5% soln.)
Physical Form Crystalline Powder and/or Crystals
Quantity 250 g
Linear Formula Na2SO4·10H2O
Fieser 01,1103
Merck Index 14,868
Solubility Information Solubility in water: soluble
Formula Weight 322.2
Percent Purity ≥99.0%
Grade Extra Pure
Packaging Plastic bottle
Chemical Name or Material Sodium sulfate decahydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.