missing translation for 'onlineSavingsMsg'
Learn More

Sodium sulfate, Puriss., meets analytical spec. of BP, Ph. Eur., USP, anhydrous, 99.0-100.5% (calc. to the dried substance), Honeywell Fluka™
BIOPHARMA

Product Code. 15651340 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
25 kg
50 kg
6 x 1 kg
6 x 2.5 kg
100 kg
Unit Size:
100kg
1kg
2.5kg
25kg
50kg
Pack of 6
Product Code. Quantity unitSize
15651340 2.5 kg 2.5kg
15641340 1 kg 1kg
15681340 6 x 1 kg Pack of 6
15691340 6 x 2.5 kg Pack of 6
15661340 25 kg 25kg
15671340 50 kg 50kg
15631340 100 kg 100kg
7 options
This item is not returnable. View return policy

Product Code. 15651340

Brand: Honeywell-Fluka 134642.5KG

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Puriss., meets analytical specification of BP, Ph. Eur., USP, anhydrous, 99.0-100.5% (calc. to the dried substance)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7757-82-6
Molecular Formula Na2O4S
Molecular Weight (g/mol) 142.04
MDL Number MFCD00003504
InChI Key PMZURENOXWZQFD-UHFFFAOYSA-L
Synonym sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1
PubChem CID 24436
ChEBI CHEBI:32149
SMILES [Na+].[Na+].[O-]S([O-])(=O)=O

Specifications

Physical Form Anhydrous
Quantity 2.5 kg
Linear Formula Na2SO4
UN Number NONH for all modes of transport
IUPAC Name disodium;sulfate
Formula Weight 142.04g/mol
Percent Purity 99 to 100.5% (calc. to the dried substance)
Grade puriss.
Density 2.68g/mL at 25°C
Chemical Name or Material Sodium Sulfate, Meets analytical specification of Ph. Eur.Bp, USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.