missing translation for 'onlineSavingsMsg'
Learn More

Sodium sulfate, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., anhydrous, ≥99.0%, Solstice

Product Code. 15663460
Change view
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
6 x 1 kg
6 x 2.5 kg
Unit Size:
1kg
2.5kg
500g
Pack of 6
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15663460 500 g 500g
15643460 1 kg 1kg
15653460 2.5 kg 2.5kg
15673460 6 x 1 kg Pack of 6
15683460 6 x 2.5 kg Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 15663460 Supplier Honeywell-Fluka Supplier No. 31481500G
  • ONLINE EXCLUSIVE PRICE
    76.00 €
    32.25 € / 500g
    Save 43.75 €
    57% Off
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., anhydrous,≥99.0%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7757-82-6
Molecular Formula Na2O4S
Molecular Weight (g/mol) 142.04
MDL Number MFCD00003504
InChI Key PMZURENOXWZQFD-UHFFFAOYSA-L
Synonym sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1
PubChem CID 24436
ChEBI CHEBI:32149
IUPAC Name disodium;sulfate
SMILES [Na+].[Na+].[O-]S([O-])(=O)=O

Specifications

Physical Form Anhydrous
Quantity 500 g
Density 2.68g/mL at 25°C
Linear Formula Na2SO4
UN Number NONH for all modes of transport
Formula Weight 142.04g/mol
Percent Purity ≥99%
Grade ACS/puriss. p.a.
Chemical Name or Material Sodium Sulfate, Reag. Ph. Eur. reag. ISO
Product Identifier
  • Sodium sulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.