missing translation for 'onlineSavingsMsg'
Learn More

Sodium sulfide, 90+%, pure, Thermo Scientific Chemicals

Product Code. 13408518 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
13408518 250 g Glass bottle 250g
10750971 10 g Glass bottle 10g
10618264 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 13408518 Supplier Thermo Scientific Acros Supplier No. 310182500
  • ONLINE EXCLUSIVE PRICE
    2015.00 €
    1954.00 € / 250g
    Save 61.00 €
    3% Off
 Restricted Item
In Stock
Añadir a la cesta
Añadir a la cesta
Request Bulk or Custom Format
Este artículo no se puede devolver. Vea la política de devoluciones
Normativa: es posible que la compra de este producto esté sujeta a restricciones y necesitemos solicitarle documentación adicional.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 1313-82-2
Molecular Formula Na2S
Molecular Weight (g/mol) 78.04
MDL Number MFCD00003498
InChI Key GRVFOGOEDUUMBP-UHFFFAOYSA-N
Synonym sodium sulfide, sodium sulfide na2s, disodium monosulfide, na2s, disodium sulphide, sodium sulfide van, hsdb 772, sodium sulfide, anhydrous dot, sodum sulfuret, na2-s
PubChem CID 237873
SMILES [Na+].[Na+].[S--]

Especificaciones

Color Beige-Yellow
Physical Form Lump/Powder
Quantity 250 g
Assay Percent Range 90% min. (Iodimetry)
Linear Formula Na2S
Merck Index 15,881
Solubility Information Solubility in water: 186g/L (20°C). Other solubilities: slightly soluble in alcohol, insoluble in ether
IUPAC Name disodium sulfanediide
Formula Weight 78.04
Percent Purity ≥90%
Grade Pure
Packaging Glass bottle
Chemical Name or Material Sodium sulfide, 90%, balance water
Mostrar más Mostrar menos

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.