missing translation for 'onlineSavingsMsg'
Learn More

Sodium sulfide, anhydrous, Thermo Scientific Chemicals

Product Code. 11364779
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11364779 100 g 100g
11354779 5 g 5g
11344779 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11364779 Supplier Thermo Scientific Alfa Aesar Supplier No. 065122.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

In organic chemicals, dyes rayon, leather and paper, in hydrometallurgy of gold ore, sulfiding oxidized lead and copper ores, and as an analytical reagentSodium sulfide is used mainly in the pulp and paper industry in the Kraft process. It serves as an oxygen scavenger agent in water treatment. Other applications include ore flotation, oil recovery, making dyes and detergents. In the leather processing industry, it is utilized for liming operation as an unhairing agent. It is also used in the textile industry as a bleaching and desulfurizing agent and as a dechlorinating agent in chemical photography for toning black and white photographs as well as metals precipitant. It plays an important role as a sulfonation and sulfomethylation agent in the chemical industry and in the production of rubber chemicals, sulfur dyes and other chemical compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1313-82-2
Molecular Formula Na2S
Molecular Weight (g/mol) 78.04
MDL Number MFCD00003498
InChI Key GRVFOGOEDUUMBP-UHFFFAOYSA-N
Synonym sodium sulfide, sodium sulfide na2s, disodium monosulfide, na2s, disodium sulphide, sodium sulfide van, hsdb 772, sodium sulfide, anhydrous dot, sodum sulfuret, na2-s
PubChem CID 237873
SMILES [Na+].[Na+].[S--]

Specifications

Melting Point 950°C
Physical Form Granules
Quantity 100 g
Solubility Information Soluble in water. Decomposes in acid. Slightly soluble in alcohol. Insoluble in etherSoluble in water. Slightly soluble in alcohol. Insoluble in ether.
IUPAC Name disodium;sulfanide
Formula Weight 78.04
Sensitivity Hygroscopic; Light sensitive
Density 1.856 g/mL
Chemical Name or Material Sodium sulfide, anhydrous

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.