missing translation for 'onlineSavingsMsg'
Learn More

Sodium sulfite, 98%, for biochemistry

Product Code. 13226329 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13226329 1 kg 1kg
13236329 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 13226329 Supplier Thermo Scientific Acros Supplier No. 447950010
  • ONLINE EXCLUSIVE PRICE
    129.00 €
    125.00 € / 1kg
    Save 4.00 €
    3% Off
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 7757-83-7 | Na2O3S | 126.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7757-83-7
Molecular Formula Na2O3S
Molecular Weight (g/mol) 126.04
InChI Key GEHJYWRUCIMESM-UHFFFAOYSA-L
Synonym sodium sulfite, disodium sulfite, sodium sulphite, sodium sulfite anhydrous, sulfurous acid, disodium salt, sulftech, natrium sulfurosum, sodiumsulfite, natriumsulfid, natriumsulfit
PubChem CID 24437
ChEBI CHEBI:86477
IUPAC Name disodium;sulfite
SMILES [O-]S(=O)[O-].[Na+].[Na+]

Specifications

Melting Point >500.0°C
Quantity 1 kg
Assay Percent Range 97.5% min. (Iodimetry)
Linear Formula Na2SO3
Merck Index 15,8811
Solubility Information (1 M in water) Clear colorless
Formula Weight 126.04
Percent Purity 98%
Grade Biochemical
Specific Gravity 2.63
Density 2.6300g/mL
Chemical Name or Material Sodium sulfite
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.