missing translation for 'onlineSavingsMsg'
Learn More

Sodium sulfite, ACS, 98.0% min

Product Code. 11326818
Change view
Click to view available options
Quantity:
2 kg
Unit Size:
2kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11326818 2 kg 2kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11326818 Supplier Thermo Scientific Alfa Aesar Supplier No. 013454.A3
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 7757-83-7 | Na{2}SO{3}

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Photographic developers, generating SO{2}, reducing agent (dyes), textile bleaching (antichlor)
TRUSTED_SUSTAINABILITY

Specifications

Melting Point >500°C (decomposition)
CAS 7757-83-7
Physical Form Solid
Quantity 2 kg
Molecular Formula Na{2}SO{3}
Percent Purity ≥98.0%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.