missing translation for 'onlineSavingsMsg'
Learn More

Sodium Sulfite, Anhydrous, ACS Reagent Grade, Ricca Chemical

Product Code. 31750011
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
31750011 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31750011 Supplier Ricca Chemical Company Supplier No. RDCS0870500B1
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

CAS 7757-83-7
Color White
CAS Min % 98.0
CAS Max % 100.0
Physical Form Crystals or Granules
Quantity 500 g
Molecular Formula Na2O3S
Synonym sodium sulfite, disodium sulfite, sodium sulphite, sodium sulfite anhydrous, sulfurous acid, disodium salt, sulftech, natrium sulfurosum, sodiumsulfite, natriumsulfid, natriumsulfit
InChI Key GEHJYWRUCIMESM-UHFFFAOYSA-L
SMILES [O-]S(=O)[O-].[Na+].[Na+]
IUPAC Name disodium;sulfite
Molecular Weight (g/mol) 126.037
PubChem CID 24437
ChEBI CHEBI:86477
Percent Purity ≥ 98%
Grade ACS
Packaging White Poly Wide Mouth Bottle
Chemical Name or Material Sodium Sulfite, Anhydrous
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.