missing translation for 'onlineSavingsMsg'
Learn More

Sodium Sulfite Anhydrous (White Crystals or Crystalline Powder), Fisher BioReagents

Product Code. 10081313
Click to view available options
Quantity:
500 g
Packaging:
Amber Glass
Unit Size:
500g
This item is not returnable. View return policy

Product Code. 10081313

Brand: Fisher BioReagents BP355500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
This sodium salt is used in buffers for molecular biology and cell culture applications.
TRUSTED_SUSTAINABILITY

Specifications

Melting Point 500°C
CAS 7757-83-7
Color White
pH 8.5 to 10
Physical Form Solid
Quantity 500 g
Molecular Formula Na2O3S
Synonym sodium sulfite, disodium sulfite, sodium sulphite, sodium sulfite anhydrous, sulfurous acid, disodium salt, sulftech, natrium sulfurosum, sodiumsulfite, natriumsulfid, natriumsulfit
InChI Key GEHJYWRUCIMESM-UHFFFAOYSA-L
SMILES [O-]S(=O)[O-].[Na+].[Na+]
IUPAC Name disodium;sulfite
Molecular Weight (g/mol) 126.037
PubChem CID 24437
ChEBI CHEBI:86477
Formula Weight 126.04
Percent Purity ≥98.0%
Identification Pass Test
Packaging Amber Glass
Density 2.630g/cm³
Chemical Name or Material Sodium Sulfite Anhydrous
Show More Show Less
Hazard Statement
  • EUH031-Contact with acids liberates toxic gas.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.