missing translation for 'onlineSavingsMsg'
Learn More

Sodium Sulfite, TRC

Product Code. 30420881
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
50 g
missing translation for 'unitSize'
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30420881 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 30420881 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCS68913550G
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sodium Sulfite
CAS 7757-83-7
Quantity 50 g
Molecular Formula Na2O3S
Synonym Sulfurous Acid Disodium Salt, Anhydrous Sodium Sulfite, Disodium Sulfite, E 221, S-WAT, Sulfurous Acid Sodium Salt, Sodium Sulfite Anhydrous
InChI Formula InChI=1S/2Na.H2O3S/c;;1-4(2)3/h;;(H2,1,2,3)/q2*+1;/p-2
SMILES [Na+].[Na+].[O-]S(=O)[O-]
IUPAC Name disodium;sulfite
Molecular Weight (g/mol) 126.04
Formula Weight 125.9364
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.