missing translation for 'onlineSavingsMsg'
Learn More

Sodium sulphite, BAKER, J.T. Baker™

Product Code. 10645812 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10645812 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10645812 Supplier JT Baker Supplier No. 1777.1000
  • ONLINE EXCLUSIVE PRICE
    53.78 €
    38.63 € / 1kg
    Save 15.15 €
    28% Off
 Restricted Item
Estimated Shipment: 07-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Laboratory Reagent

Specifications

Melting Point >500°C
CAS 7757-83-7
Quantity 1 kg
Assay Percent Range min. 95%
Molecular Formula H14Na2O10S
MDL Number MFCD00003503
InChI Key WMJRPJZQQSSDBU-UHFFFAOYSA-L
SMILES O.O.O.O.O.O.O.[Na+].[Na+].[O-]S([O-])=O
IUPAC Name disodium sulfite
Molecular Weight (g/mol) 252.14
Density 2.63 g/cm³ (20°C)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.