missing translation for 'onlineSavingsMsg'
Learn More

Sodium Tartrate Dihydrate, ACS Reagent Grade, Ricca Chemical

Product Code. 31749380
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31749380 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31749380 Supplier Ricca Chemical Company Supplier No. RDCS0890500B1
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 6106-24-7
CAS Min % 98.0
CAS Max % 100.0
Physical Form Fine Crystals or Crystalline Powder
Quantity 500 g
Packaging White Poly Wide Mouth Bottle
Molecular Formula C4H8Na2O8
MDL Number MFCD00150035
Synonym unii-dia7c37aow, disodium tartrate dihydrate, disodium l-+-tartrate dihydrate, dia7c37aow, disodium +-tartrate dihydrate, sodium l-+-tartrate dihydrate, butanedioic acid, 2,3-dihydroxy-2r,3r-, disodium salt, dihydrate, disodium tartarate dihydrate, sodium tartrate nf, sodium tartrate dihydrate mi
InChI Key FGJLAJMGHXGFDE-UDMGOBQRNA-L
SMILES O.O.[Na+].[Na+].O[C@H]([C@@H](O)C([O-])=O)C([O-])=O
IUPAC Name disodium (2R,3R)-2,3-dihydroxybutanedioate dihydrate
Molecular Weight (g/mol) 230.08
PubChem CID 131855972
Percent Purity 99 to 101%
Grade ACS
Chemical Name or Material Sodium Tartrate, Dihydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.