missing translation for 'onlineSavingsMsg'
Learn More

Sodium Tartrate Dihydrate, Certified AR for Analysis, Fisher Chemical

Product Code. 10624932 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Plastic powder jar
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10624932 500 g Plastic powder jar 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10624932 Supplier Fisher Chemical Supplier No. S/6922/53
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 6106-24-7
Color White
pH 7 to 9
Physical Form Powder/Solid
Quantity 500 g
Molecular Formula C4H8Na2O8
MDL Number MFCD00150035
Synonym unii-dia7c37aow, disodium tartrate dihydrate, disodium l-+-tartrate dihydrate, dia7c37aow, disodium +-tartrate dihydrate, sodium l-+-tartrate dihydrate, butanedioic acid, 2,3-dihydroxy-2r,3r-, disodium salt, dihydrate, disodium tartarate dihydrate, sodium tartrate nf, sodium tartrate dihydrate mi
InChI Key FGJLAJMGHXGFDE-UDMGOBQRNA-L
SMILES O.O.[Na+].[Na+].O[C@H]([C@@H](O)C([O-])=O)C([O-])=O
IUPAC Name disodium (2R,3R)-2,3-dihydroxybutanedioate dihydrate
Molecular Weight (g/mol) 230.08
PubChem CID 131855972
Formula Weight 230.08g/mol
Packaging Plastic powder jar
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.