missing translation for 'onlineSavingsMsg'
Learn More

Sodium tellurate(VI) hydrate, Te 45% min

Product Code. 11304889
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11304889 50 g 50g
11314889 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11304889 Supplier Thermo Scientific Alfa Aesar Supplier No. 075106.18
 Restricted Item
Estimated Shipment: 08-12-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

CAS: 10101-83-4 ∣ Na2O4Te ∣ 237.58 g/mol

Sodium tellurite is an intermediate in the extraction of the element, tellurium; it is a product obtained from anode slimes and is a precursor to tellurium. Solutions of sodium tellurite are used for black or blue-black coatings on iron, steel, aluminum, and copper.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 10101-83-4
Molecular Formula Na2O4Te
Molecular Weight (g/mol) 237.58
MDL Number MFCD00014242
InChI Key XERQTZLDFHNZIC-UHFFFAOYSA-L
Synonym sodium tellurate, unii-7kar02f0r0, natriumtellurat, 2na.o4te, disodium tellurate, telluric acid h2teo4 , sodium salt 1:2, telluric acid h2teo4 ,disodium salt, dihydrate 8ci,9ci
PubChem CID 13652393
SMILES [Na+].[Na+].[O-][Te]([O-])(=O)=O

Spezifikation

Melting Point (decomposition)
Physical Form Crystalline
Quantity 50 g
Merck Index 14,8686
Solubility Information Soluble in water.
IUPAC Name disodium;tellurate
Formula Weight 237.58 (Anhydrous)
Grade Ungraded
Concentration or Composition (by Analyte or Components) Te 45% min
Chemical Name or Material Sodium tellurate (VI) hydrate

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.