missing translation for 'onlineSavingsMsg'
Learn More

Sodium tert-butoxide, 99%, pure

Product Code. 10247350 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic Drum
Unit Size:
100g
2.5kg
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10247350 100 g Glass bottle 100g
10728362 5 g Glass bottle 5g
10723924 500 g Glass bottle 500g
11905821 2.5 kg Plastic Drum 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10247350 Supplier Thermo Scientific Acros Supplier No. 252551000
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 865-48-5 | C4H9NaO | 96.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 865-48-5
Molecular Formula C4H9NaO
Molecular Weight (g/mol) 96.1
MDL Number MFCD00040575
InChI Key MFRIHAYPQRLWNB-UHFFFAOYSA-N
Synonym sodium tert-butoxide, sodium 2-methylpropan-2-olate, sodium t-butoxide, sodium-t-butoxide, t-butoxy sodium, tert-butoxysodium, naotbu, tbuona, 2-propanol, 2-methyl-, sodium salt, sodium-tert-butoxide
PubChem CID 23676156
IUPAC Name sodium;2-methylpropan-2-olate
SMILES CC(C)(C)[O-].[Na+]

Specifications

Melting Point 180.0°C
Flash Point 12°C
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula NaOC(CH3)3
Quantity 100 g
Fieser 16,55
Solubility Information Solubility in water: reacts.
Formula Weight 96.1
Percent Purity 99%
Grade Pure
Chemical Name or Material Sodium tert-butoxide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Sodium tert-butoxide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable solid Category 1
  • Self-heating substance Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H251-Self-heating: may catch fire.
  • H228-Flammable solid.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.