missing translation for 'onlineSavingsMsg'
Learn More

Sodium tetraborate decahydrate, 99.5+%, ACS reagent

Product Code. 10269370 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
1 kg
2.5 kg
Packaging:
Plastic bottle
Unit Size:
1kg
2.5kg
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10269370 1 kg Plastic bottle 1kg
10577562 25 g Plastic bottle 25g
10286843 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10269370 Supplier Thermo Scientific Acros Supplier No. 419450010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1303-96-4 | B4Na2O7·10H2O | 381.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1303-96-4
Molecular Formula B4Na2O7·10H2O
Molecular Weight (g/mol) 381.36
MDL Number MFCD00149193
Synonym Borax, Sodium borate decahydrate

Specifications

Melting Point 75.0°C
Color White
pH 9.15 to 9.20 (0.01M soln. at 25°C)
Physical Form Crystalline Powder
Quantity 1 kg
Assay Percent Range 99.5+%
Linear Formula Na2B4O7·10H2O
Fieser 04,461
Merck Index 15, 8726
Solubility Information Solubility in water: 60g/L (20°C). Other solubilities: soluble in glycerol, insoluble in alcohol
Formula Weight 381.36
Percent Purity ≥99.5%
Grade ACS Reagent
Packaging Plastic bottle
Chemical Name or Material Sodium tetraborate decahydrate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Sodium tetraborate decahydrate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 1B
Hazard Statement
  • H319-Causes serious eye irritation.
  • H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.