missing translation for 'onlineSavingsMsg'
Learn More

Sodium tetraborate decahydrate, 99+%

Product Code. 11493237
Change view
Click to view available options
Quantity:
10,000 g
2500 g
500 g
Unit Size:
10000g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11493237 500 g 500g
11403247 2500 g 2500g
11413247 10,000 g 10000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11493237 Supplier Thermo Scientific Alfa Aesar Supplier No. A16176.36
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1303-96-4 ∣ B4Na2O7·10H2O

It is used in detergents, cosmetics, and enamel glazes. It is likewise employed to prepare buffer solutions in biochemistry, as a fire retardant, anti-fungal compound, in the manufacture of fiberglass, as a flux in metallurgy, neutron-capture shields for radioactive sources, a texturing agent in cooking, and as a harbinger for other boron compounds. The molten borax bath is useful for carbide coating process on metals. It is applied as an efficient metal-free catalyst for Hetero-Michael reactions in an aqueous medium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications

It is used in detergents, cosmetics, and enamel glazes. It is likewise employed to prepare buffer solutions in biochemistry, as a fire retardant, anti-fungal compound, in the manufacture of fiberglass, as a flux in metallurgy, neutron-capture shields for radioactive sources, a texturing agent in cooking, and as a harbinger for other boron compounds. The molten borax bath is useful for carbide coating process on metals. It is applied as an efficient metal-free catalyst for Hetero-Michael reactions in an aqueous medium.

Solubility

Soluble in water, glycerol and ethanol. Insoluble in acids

Notes

Incompatible with strong oxidizing agents and strong acids.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1303-96-4
Molecular Formula B4Na2O7·10H2O
MDL Number MFCD00149193
Synonym Borax; Sodium borate (tetra) decahydrate

Spezifikation

Melting Point 75°C
pH 9.24
Physical Form Crystalline Powder and/or Crystals
Quantity 500 g
Odor Odorless
Density 1.73
Linear Formula Na2B4O7·10H2O
Merck Index 14,8590
Solubility Information Soluble in water,glycerol and ethanol. Insoluble in acids
Molecular Weight (g/mol) 381.36
Formula Weight 381.37 (201.22 Anhydrous)
Percent Purity ≥99%
Grade Ungraded
Chemical Name or Material Sodium tetraborate decahydrate
Mehr anzeigen Weniger anzeigen
compliance-icons compliance-icons
Produktidentifikator
  • Sodium tetraborate decahydrate
Signalwort
  • Danger
Gefahrenkategorie
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 1B
Gefahrenhinweis
  • H319-Causes serious eye irritation.
  • H360FD-May damage fertility. May damage the unborn child.
Sicherheitshinweis
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Ergänzende Informationen
  • MIXTURE LIST-Contain: borax

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.