missing translation for 'onlineSavingsMsg'
Learn More

Sodium tetraborate decahydrate, Puriss., meets analytical spec. of BP, NF, Ph. Eur., 99.0-103.0%, Honeywell Fluka™
BIOPHARMA

Product Code. 15621070 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
2.5 kg
25 kg
6 x 2.5 kg
Packaging:
Paper / Poly Bag
Poly bottle
Unit Size:
2.5kg
25kg
Pack of 6
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15621070 25 kg Paper / Poly Bag 25kg
15611070 2.5 kg Poly bottle 2.5kg
15631070 6 x 2.5 kg Poly bottle Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15621070 Supplier Honeywell-Fluka Supplier No. 1162525KG
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Puriss., meets analytical specification of BP, NF, Ph. Eur., 99.0-103.0%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1303-96-4
MDL Number MFCD00149193
Synonym Borax decahydrate, Sodium borate decahydrate

Specifications

Boiling Point 1,575°C
CAS Max % 100.0000%
Quantity 25 kg
Assay Percent Range 99 to 103.0%
Linear Formula Na2B4O7 · 10H2O
Formula Weight 381.37
Grade puriss.
Packaging Paper / Poly Bag
Density 1.73 g/mL at 25 °C(lit.)
Chemical Name or Material Sodium tetraborate decahydrate, Puriss., meets analytical specification of BP, NF, Ph. Eur., 99.0-103.0%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.