missing translation for 'onlineSavingsMsg'
Learn More

Sodium Tetraborate Decahydrate, puriss. p.a. ACS Reagent, Solstice

Product Code. 15663450 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
5 kg
25 kg
Unit Size:
100g
1kg
25kg
500g
5kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15663450 5 kg 5kg
15623450 100 g 100g
15653450 500 g 500g
15633450 1 kg 1kg
15643450 25 kg 25kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 15663450 Supplier Honeywell-Fluka Supplier No. 314575KG
  • ONLINE EXCLUSIVE PRICE
    371.00 €
    335.38 € / 5kg
    Save 35.62 €
    9% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Puriss. p.a., ACS Reagent, Reagent ISO, buffer substance,≥99.5%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1303-96-4
MDL Number MFCD00149193

Specifications

Physical Form Crystals
Quantity 5 kg
Linear Formula Na2B4O7 · 10H2O
UN Number NONH for all modes of transport
Molecular Weight (g/mol) 381.37
Formula Weight 381.37g/mol
Percent Purity ≥99.5%
Grade ACS/puriss. p.a.
Density 1.73g/mL at 25°C
Chemical Name or Material Sodium Tetraborate Decahydrate, Reag. ISO
compliance-icons
Product Identifier
  • Sodium tetraborate decahydrate
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1B
Hazard Statement
  • H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.