missing translation for 'onlineSavingsMsg'
Learn More

Sodium Tetraborate, TRC

Product Code. 30422813
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30422813 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30422813 Supplier Toronto Research Chemicals Supplier No. TRCS673095100G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Disodium tetraborate
CAS 1330-43-4
Quantity 100 g
Molecular Formula B4 O7 . 2Na
Synonym Sodium tetraborate, Boron sodium oxide (B4Na2O7) (9CI','ACI), Boric acid (H2B4O7)','disodium salt (8CI), Sodium tetraborate (Na2B4O7) (7CI), Anhydrous borax, Borax glass, DC Dehybor, Dehybor, Ecoboron PRO, Etibor 68, Fireless B, Fireless B Liquid, FR 28, FR 28 (borate), Fused Borax, Granubor, Rasorite 65, Sodium biborate, Sodium borate, Sodium boron oxide (Na2B4O7), Spraybor
InChI Formula InChI=1S/B4O7.2Na/c5-1-7-3-9-2(6)10-4(8-1)11-3;;/q-2;2*+1
SMILES [Na+].[Na+].[O-]B1OB2OB([O-])OB(O1)O2
IUPAC Name disodium;3,7-dioxido-2,4,6,8,9-pentaoxa-1,3,5,7-tetraborabicyclo[3.3.1]nonane
Molecular Weight (g/mol) 201.22
Formula Weight 201.9812
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.