missing translation for 'onlineSavingsMsg'
Learn More

Sodium tetrachloroaurate hydrate, 98%

Product Code. 12614997 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 12614997

Brand: Thermo Scientific Acros 396270050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 29156-65-8 | AuCl4Na | 361.76 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29156-65-8
Molecular Formula AuCl4Na
Molecular Weight (g/mol) 361.76
MDL Number MFCD00149161
InChI Key IXPWAPCEBHEFOV-UHFFFAOYSA-J
Synonym sodium tetrachloroaurate iii hydrate, aucl4.na.h2o, sodium tetrachloroaurate 1--water 1/1/1
PubChem CID 18410391
IUPAC Name sodium;tetrachlorogold(1-);hydrate
SMILES [Na+].Cl[Au-](Cl)(Cl)Cl

Specifications

Color Brown to Yellow
Physical Form Lump/Powder
Quantity 5 g
Assay Percent Range 48.3 to 50.3% (Au)
Linear Formula NaAuCl4·xH2O
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol and ether
Formula Weight 361.77
Percent Purity 98%
Packaging Glass bottle
Chemical Name or Material Sodium tetrachloroaurate hydrate
compliance-icons compliance-icons
Product Identifier
  • Sodium tetrachloroaurate hydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H290-May be corrosive to metals.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.