missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium tetrachloroaurate hydrate, 98%
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Brown to Yellow |
| Physical Form | Lump/Powder |
| Quantity | 1 g |
| Assay Percent Range | 48.3 to 50.3% (Au) |
| Linear Formula | NaAuCl4·xH2O |
| Solubility Information | Solubility in water: soluble. Other solubilities: soluble in alcohol and ether |
| Formula Weight | 361.77 |
| Percent Purity | 98% |
| Packaging | Glass bottle |
| Chemical Name or Material | Sodium tetrachloroaurate hydrate |
Safety and Handling
Product Identifier
- Sodium tetrachloroaurate hydrate
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
- H290-May be corrosive to metals.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction