missing translation for 'onlineSavingsMsg'
Learn More

Sodium tetraethylborate, 97%, pure, Thermo Scientific Chemicals

Product Code. 10679293 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10679293 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Product Code. 10679293 Supplier Thermo Scientific Acros Supplier No. 363880050
  • ONLINE EXCLUSIVE PRICE
    424.00 €
    397.00 € / 5g
    Save 27.00 €
    6% Off
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.

CAS: 15523-24-7 | Molecular Formula: C8H20BNa | Molecular Weight: 150.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15523-24-7
Molecular Formula C8H20BNa
Molecular Weight (g/mol) 150.04
MDL Number MFCD00061547
InChI Key SZSBMTRYJRHYNI-UHFFFAOYSA-N
Synonym sodium tetraethylborate, sodium tetraethylboranuide, borate 1-,tetraethyl-, sodium 1:1, sodium tetraethylborate 1-, et4bna, sodiotetraethylboron v, borate 1-, tetraethyl-, sodium, sodium tetraethylborate 1g
PubChem CID 23681030
IUPAC Name sodium;tetraethylboranuide
SMILES [B-](CC)(CC)(CC)CC.[Na+]

Specifications

Melting Point 140.0°C to 142.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: reacts
Formula Weight 150.04
Percent Purity 97%
Grade Pure
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Sodium tetraethylborate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.