missing translation for 'onlineSavingsMsg'
Learn More

Sodium thiocyanate, 96%

Product Code. 11473834
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11473834 1000 g 1000g
11483834 250 g 250g
11493834 5000 g 5000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11473834 Supplier Thermo Scientific Alfa Aesar Supplier No. A11867.0B
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 540-72-7 | CNNaS | 81.07 g/mol

A compound used to convert alkyl halides into the corresponding alkylthiocyanates. Undergoes nucleophilic substitution reactions with phase-transfer catalysts. Silver thiocyanate may be used as well, the precipitation of insoluble silver halides help simplify workup. Treatment of isopropyl bromide with sodium thiocyanate in a hot ethanolic solution affords isopropyl thiocyanate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A compound used to convert alkyl halides into the corresponding alkylthiocyanates. Undergoes nucleophilic substitution reactions with phase-transfer catalysts. Silver thiocyanate may be used as well, the precipitation of insoluble silver halides help simplify workup. Treatment of isopropyl bromide with sodium thiocyanate in a hot ethanolic solution affords isopropyl thiocyanate.

Solubility
Soluble in water (1250g/L), alcohol, and acetone.

Notes
Hygroscopic, protect from humidity and water. Store in cool, dry conditions in well sealed container.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 540-72-7
Molecular Formula CNNaS
Molecular Weight (g/mol) 81.07
MDL Number MFCD00011123
InChI Key VGTPCRGMBIAPIM-UHFFFAOYSA-M
Synonym sodium thiocyanate, sodium rhodanate, sodium sulfocyanate, sodium rhodanide, thiocyanic acid, sodium salt, thiocyanate sodium, scyan, sodium isothiocyanate, nascn, haimased
PubChem CID 516871
ChEBI CHEBI:30952
IUPAC Name sodium;thiocyanate
SMILES [Na+].[S-]C#N

Specifications

Melting Point 287°C
Density 1.74
pH 6 to 8
Odor Odorless
Quantity 1000 g
Beilstein 3594965
Sensitivity Hygroscopic; Light sensitive
Merck Index 14,9327
Solubility Information Soluble in water (1250g/L),alcohol,and acetone.
Formula Weight 81.07
Percent Purity 96%
Chemical Name or Material Sodium thiocyanate
Show More Show Less
compliance-icons
Product Identifier
  • Sodium thiocyanate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.
  • EUH032-Contact with acids liberates very toxic gas.
Precautionary Statement
  • P233-Keep container tightly closed.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Supplemental information
  • MIXTURE LIST-Contain: Sodium thiocyanate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.